Rat Recombinant IFNG/Interferon Gamma Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80234-R02H-100
103666-516EA 250.48 USD
103666-516
Rat Recombinant IFNG/Interferon Gamma Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat IFNG (P01581) (Met 1-Cys 156) was fused with the Fc region of human IgG1 at the C-terminus.