Rat Recombinant IL20RB Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80187-R08H-100
103666-428EA 300.98 USD
103666-428
Rat Recombinant IL20RB Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat IL20RB (B8K1X9) (Met1-Pro227) was expressed, fused with a polyhistidine tag at the C-terminus.