Rat Recombinant BAFFR/TNFRSF13C Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80171-R02H-100
103666-398EA 300.98 USD
103666-398
Rat Recombinant BAFFR/TNFRSF13C Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat TNFRSF13C (XP_576316.2) (Ser10-Ala71) was expressed with the Fc region of human IgG1 at the C-terminus.