Rat Recombinant VEGF-D/VEGFD/FIGF Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80104-R01H-50
103668-622EA 300.98 USD
103668-622
Rat Recombinant VEGF-D/VEGFD/FIGF Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat FIGF (NP_113949.1)(Phe94-Arg210) was expressed, fused with the Fc region of human IgG1 at the N-terminus.