Rat Recombinant FAS/CD95/APO-1/TNFRSF6 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80043-R08H-100
103666-286EA 300.98 USD
103666-286
Rat Recombinant FAS/CD95/APO-1/TNFRSF6 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat FAS (NP_631933.2) extracellular domain (Met 1-Lys 170) was fused with a polyhistidine tag at the C-terminus.