Canine Recombinant RBP4 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
70006-D08H-100
103666-066EA 300.98 USD
103666-066
Canine Recombinant RBP4 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine RBP4 (F1PQI4) (Met1-Leu201) was expressed with a C-terminal polyhistidine tag.