Canine Recombinant KIM-1/TIM1/HAVCR1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
70001-D04H-200
103666-056EA 452.48 USD
103666-056
Canine Recombinant KIM-1/TIM1/HAVCR1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of canine HAVCR1 (XP_854888.1) (Tyr21-Ser160) was fused with the Fc region of mouse IgG at the N-terminus.