Mouse Recombinant Neuropilin-2/NRP2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
57465-M08H-100 57465-M08H-20
103666-004EA 351.48 USD
103666-004 103668-874
Mouse Recombinant Neuropilin-2/NRP2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse NRP2 (NP_001070871.1) (Met1-Pro864) was expressed with a polyhistidine tag at the C-terminus.