Mouse Recombinant VEGFR2/Flk-1/CD309/KDR Protein (His Tag) , Biotinylated (from HEK293 Cells)
Supplier: Sino Biological
50998-M08H-B-20
103634-042EA 179.78 USD
103634-042
Mouse Recombinant VEGFR2/Flk-1/CD309/KDR Protein (His Tag) , Biotinylated (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse KDR (P35918-1) extracellular domain (Met1-Glu762) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.