Mouse Recombinant CD63/Tspan-30/Tetraspanin-30 Protein (Fc Tag, ECD) (from HEK293 Cells)
Supplier: Sino Biological
50557-M01H-100 50557-M01H-20
103665-144EA 351.48 USD
103665-144 103667-678
Mouse Recombinant CD63/Tspan-30/Tetraspanin-30 Protein (Fc Tag, ECD) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse Cd63 (NP_031679.1) (Ala103-Ile203) was expressed with the Fc region of human IgG1 at the N-terminus.