Mouse Recombinant CD19/Leu-12 Protein (ECD, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50510-M08H-50
103667-560EA 250.48 USD
103667-560
Mouse Recombinant CD19/Leu-12 Protein (ECD, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse CD19 (NP_033974.2) (Met1-Gly287) was expressed, with a polyhistidine tag at the C-terminus.