Mouse Recombinant CD180/RP105 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50491-M08H-50 50491-M08H-100
103665-074EA 300.98 USD
103665-074 103667-542
Mouse Recombinant CD180/RP105 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CD180 (NP_032559.2) extracellular domain (Met 1-Ser 626) was expressed, fused with a polyhistidine tag at the C-terminus.