Mouse Recombinant PRLR/Prolactin Receptor Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50457-M03H-50 50457-M03H-100
103667-474EA 191.35 USD
103667-474 103664-446
Mouse Recombinant PRLR/Prolactin Receptor Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse PRLR (NP_035299.4) (Met 1-Asp 229) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.