Mouse Recombinant CCL2/MCP-1/MCP1 Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
50368-M08B-50 50368-M08B-20
103667-308EA 250.48 USD
103667-308 103667-306
Mouse Recombinant CCL2/MCP-1/MCP1 Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CCL2 (NP_035463.1) (Met1-Asn148) was expressed with a C-terminal polyhistidine tag.