Mouse Recombinant TrkC/NTRK3 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50320-M08H-100 50320-M08H-50
103667-240EA 191.35 USD
103667-240 103664-888
Mouse Recombinant TrkC/NTRK3 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse TrkC (NP_032772.3) (Met 1-Thr 429) was expressed, with a C-terminal polyhistidine tag.