Mouse Recombinant SRC Kinase/Proto-oncogene c-Src (His & GST Tag) Protein (from Baculovirus Insect Cells)
Supplier: Sino Biological
50311-M20B-50 50311-M20B-20
103667-214EA 212.85 USD
103667-214 103667-216
Mouse Recombinant SRC Kinase/Proto-oncogene c-Src (His & GST Tag) Protein (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the mouse SRC (NP_001020566.111) (Met 1-Leu 535) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.