Mouse Recombinant DR6/TNFRSF21 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50263-M08H-100 50263-M08H-50
103664-822EA 300.98 USD
103664-822 103667-090
Mouse Recombinant DR6/TNFRSF21 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse TNFRSF21 (NP_848704.1) extracellular domain (Met 1-His 349) was fused with a polyhistidine tag at the C-terminus.