Mouse Recombinant TNFRSF19/TROY Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50148-M08H-100 50148-M08H-200
103664-674EA 320.35 USD
103664-674 103664-672
Mouse Recombinant TNFRSF19/TROY Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Leu 170) of mouse TNFRSF19 (NP_001157627.1) was fused with a polyhistidine tag at the C-terminus.