Mouse Recombinant PD-L1/B7-H1/CD274 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50010-M02H-100
103664-372EA 179.78 USD
103664-372
Mouse Recombinant PD-L1/B7-H1/CD274 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse Cd274 (NP_068693.1) (Met1-His239) was expressed with the Fc region of human IgG1 at the C-terminus.