Influenza A H1N1 (A/Baylor/11515/1982) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40728-V07H-100 40728-V07H-100
103887-458EA 401.98 USD
103887-458 CA76477-858
Influenza A H1N1 (A/Baylor/11515/1982) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the influenza A virus (A/Baylor/11515/1982 (H1N1)) neuraminidase (ABD95342) (His36-Lys470), termed as NA, was fused with a N-terminal polyhistidine tag.