Influenza A H1N1 (A/Tawain/01/1986) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40712-V08H-100 40712-V08H-100
103887-448EA 351.48 USD
103887-448 CA76477-848
Influenza A H1N1 (A/Tawain/01/1986) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Tawain/01/1986 (H1N1)) hemagglutinin (ABF21274.1) (Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.