Influenza A H1N2 (A/swine/Hong Kong/4083/2011) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40676-V08H-100 40676-V08H-100
103887-400EA 351.48 USD
103887-400 CA76477-800
Influenza A H1N2 (A/swine/Hong Kong/4083/2011) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/Hong Kong/4083/2011 (H1N2)) hemagglutinin (AIE50626) (Ala17-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.