Influenza A H1N1 (A/Michigan/45/2015) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40568-V07H-100 40568-V07H-100
103918-610EA 401.98 USD
CA76487-742 103918-610
Influenza A H1N1 (A/Michigan/45/2015) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Michigan/45/2015 (H1N1pdm09)) neuraminidase (translated amino acids of EPI859651) (His36-Lys469) was expressed with a polyhistidine tag at the N-terminus.