Dengue virus (DENV) Recombinant (type 4, strain Philippines/H241/1956) E/Envelope Protein (aa 579-679, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40533-V08H-100 40533-V08H-100
103906-334EA 351.48 USD
CA76479-994 103906-334
Dengue virus (DENV) Recombinant (type 4, strain Philippines/H241/1956) E/Envelope Protein (aa 579-679, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the envelope protein of Dengue virus (DENV)(type 4, strain Philippines/H241/1956) Envelope Protein (AAX48017.1) (Thr579-Lys679) was expressed with a polyhistidine tag at the C-terminus.