Influenza A H5N2 (A/chicken/Iowa/04-20/2015) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40518-V08B-100
103664-302EA 351.48 USD
103664-302
Influenza A H5N2 (A/chicken/Iowa/04-20/2015) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/chicken/Iowa/04-20/2015)(H5N2)) hemagglutinin (AKG52687.1) (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.