Influenza A H6N6 (A/duck/Eastern China/11/2009) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40399-V08H1-100
103633-286EA 351.48 USD
103633-286
Influenza A H6N6 (A/duck/Eastern China/11/2009) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Eastern China/11/2009(H6N6)) hemagglutinin (AEF59343.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.