Influenza A H5N8 (A/duck/Zhejiang/W24/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40387-V08H1-100
103634-020EA 351.48 USD
103634-020
Influenza A H5N8 (A/duck/Zhejiang/W24/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Zhejiang/W24/2013(H5N8)) hemagglutinin (AHM24057.1)(Met1-Lys341), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.