Influenza A H10N3 (A/duck/Hunan/S11205/2012) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40360-V08H1-100
103633-266EA 351.48 USD
103633-266
Influenza A H10N3 (A/duck/Hunan/S11205/2012) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Hunan/S11205/2012 (H10N3)) hemagglutinin (AGO87051.1) (Met1-Arg340), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.