HIV-1 Recombinant gp160 (gp120 subunit) (group N, strain 06CM-U14296) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40255-V08H-100
103663-982EA 351.48 USD
103663-982
HIV-1 Recombinant gp160 (gp120 subunit) (group N, strain 06CM-U14296) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the HIV-1 Envelope glycoprotein gp160 protein (Trp32-Arg504) was expressed with a polyhistidine tag at the C-terminus.