Influenza B (B/Massachusetts/03/2010) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40191-V08H1-100
103633-148EA 351.48 USD
103633-148
Influenza B (B/Massachusetts/03/2010) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza B virus (B/Massachusetts/03/2010) hemagglutinin (AGI64446.1) (Met1-Arg362), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.