Influenza A H9N2 (A/Hong Kong/3239/2008) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40178-V08H-100
103663-906EA 351.48 USD
103663-906
Influenza A H9N2 (A/Hong Kong/3239/2008) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hong Kong/3239/2008 (H9N2)) hemagglutinin (ADC41863.1) (Met1-Arg338), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.