Influenza A H1N1 (A/USSR/90/1977) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40134-V08H1-100
103632-924EA 351.48 USD
103632-924
Influenza A H1N1 (A/USSR/90/1977) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/USSR/90/1977(H1N1)) hemagglutinin (P03453) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.