MERS-CoV Recombinant Spike/S1 Protein (S1 Subunit, aa 1-725, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40069-V08H-100
103663-610EA 452.48 USD
103663-610
MERS-CoV Recombinant Spike/S1 Protein (S1 Subunit, aa 1-725, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the MERS-CoV spike protein S1 Subunit (AFS88936.1)(Met1-Glu725) was fused with a polyhistidine tag at the C-terminus.