Influenza A H3N2 Recombinant Neuraminidase/NA (N294S mutation) (Active) Protein (from HEK293 Cells)
Supplier: Sino Biological
40017-VNAHC2-80 40017-VNAHC2-500
103634-068EA 292.9 USD
103634-068 103633-912
Influenza A H3N2 Recombinant Neuraminidase/NA (N294S mutation) (Active) Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Babol/36/2005 (H3N2)) neuraminidase (ACN50232.1) (His 36-Pro 459) was expressed, the cell lysates are collected, and bio-activity was tested. There is an amino acid change from Asparagine to Serine (N294S mutation) in NA / Neuraminidase.