Influenza B (B/Brisbane/60/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40016-V08H-100
103663-498EA 351.48 USD
103663-498
Influenza B (B/Brisbane/60/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Ala 556) of the influenza B hemagglutinin (B/Brisbane/60/2008)) (ACN29380.1) was fused with a polyhistidine tag at the C-terminus.