Influenza A H5N1 (A/Duck/Hong Kong/p46/97) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40001-V08H-100
103663-448EA 351.48 USD
103663-448
Influenza A H5N1 (A/Duck/Hong Kong/p46/97) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Gln539) of the influenza hemagglutinin (A/Duck/Hong Kong/p46/97 (H5N1)) (AAF02306.1) (HA1+HA2, uncleaved), cleavage site mutated (RRRKKR-TETR) was fused with a polyhistidine tag at the C-terminus.