Human Recombinant LAG3/CD223/Lymphocyte activation gene 3 Protein (ECD,Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
16498-H05H-50
103614-988EA 179.78 USD
103614-988
Human Recombinant LAG3/CD223/Lymphocyte activation gene 3 Protein (ECD,Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LAG3 (NP_002277.4) (Met1-Leu450) was expressed with the Fc region of mouse IgG1 at the C-terminus.