Human Recombinant GYPA/CD235a/Glycophorin A Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
16018-H08H-100 16018-H08H-200
103663-292EA 300.98 USD
103663-292 103663-294
Human Recombinant GYPA/CD235a/Glycophorin A Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human GYPA (P02724) (Met1-Glu91) was expressed with a C-terminal polyhistidine tag.