Human Recombinant MANF/ARMET Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
15548-H02H-100 15548-H02H-5 15548-H02H-20
103663-128EA 300.98 USD
103663-128 103614-496 103677-722
Human Recombinant MANF/ARMET Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human MANF (NP_006001.4) (Met1-Leu182) was expressed with the Fc region of human IgG1 at the C-terminus.