Human Recombinant CD81/TAPA-1 Protein (His Tag, ECD) (from HEK293 Cells)
Supplier: Sino Biological
14244-H07H-20 14244-H07H-100
103614-340EA 148.35 USD
103614-340 103623-632
Human Recombinant CD81/TAPA-1 Protein (His Tag, ECD) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CD81 (NP_004347.1) (Phe113-Lys201) was expressed with a polyhistidine tag at the N-terminus.