Human Recombinant CD45 Protein (ECD, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
14197-H08H-50 14197-H08H-50
103918-868EA 199.98 USD
103918-868 CA76488-000
Human Recombinant CD45 Protein (ECD, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CD45 (NP_002829.3) (Gln26-Lys575) was expressed with a polyhistidine tag at the C-terminus.