Human Recombinant EphA2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13926-H08H-100 13926-H08H-200
103623-524EA 320.35 USD
103623-524 103623-114
Human Recombinant EphA2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human EPHA2 (P29317) (Met1-Asn534) was expressed with a polyhistidine tag at the C-terminus.