Human Recombinant FUCA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13893-H08H-20 13893-H08H-10
103614-246EA 159.58 USD
103614-246 103614-244
Human Recombinant FUCA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human FUCA1 (P04066) (Gln32-Lys466) was expressed with a polyhistidine tag at the C-terminus.