Human Recombinant Interferon alpha 2/IFNA2 Protein (from Yeast)
Supplier: Sino Biological
13833-HNAY-200 13833-HNAY-100
103623-086EA 351.48 USD
103623-086 103623-084
Human Recombinant Interferon alpha 2/IFNA2 Protein (from Yeast)
Proteins and Peptides
A DNA sequence encoding the human IFNA2 (P01563.1) (Cys24-Glu188) was expressed.