Human Recombinant PAM/Peptidylglycine alpha‑Amidating Monooxygenase Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13624-H08H-10 13624-H08H-50
103613-542EA 191.35 USD
103613-542 103613-544
Human Recombinant PAM/Peptidylglycine alpha‑Amidating Monooxygenase Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human PAM (NP_620176.1) (Met1-Ser866) was expressed with a polyhistidine tag at the C-terminus.