Human Recombinant FGL1 Protein (mFc Tag) (from HEK293 Cells)
Supplier: Sino Biological
13484-H38H-100 13484-H38H-1 13484-H38H-1 13484-H38H-100
103885-840EA 2270.48 USD
CA76475-946 103885-840 103885-846 CA76475-952
Human Recombinant FGL1 Protein (mFc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human FGL1 (NP_004458.3) (Met1-Ile312) was expressed with the Fc region of mouse IgG2a at the C-terminus.