Human Recombinant CLEC3B/Tetranectin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
12735-H08H-100 12735-H08H-50
103616-088EA 191.35 USD
103616-088 103623-208
Human Recombinant CLEC3B/Tetranectin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the second extracellular domain of human CLEC3B (AAX37102.1) (Met 1-Val 202) was fused with a polyhistidine tag at the C-terminus.