Human Recombinant KRAS/K-Ras (Q61H) Protein (His Tag) (from E. coli)
Supplier: Sino Biological
12259-H07E-20 12259-H07E-50
103615-808EA 148.35 USD
103615-808 103615-810
Human Recombinant KRAS/K-Ras (Q61H) Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human KRAS (P01116-2) (Thr2-Cys185, with natural variant Gln 61 His ) was expressed with a polyhistide tag at the N-terminus.