Human Recombinant PCNA Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
12131-H07B-20 12131-H07B-100
103622-984EA 351.48 USD
103622-984 103613-030
Human Recombinant PCNA Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human PCNA (P12004) (Met 1-Ser 261) was expressed, with a polyhistidine tag at the N-terminus.