Human Recombinant PRNP/Prion Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
12070-H02H-100 12070-H02H-20
103612-990EA 148.35 USD
103612-990 103622-948
Human Recombinant PRNP/Prion Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human PRNP (NP_000302.1) (Met1-Gly229) was expressed with the Fc region of human IgG1 at the C-terminus.